siRNA Details
| Gene Symbol: 9130401L11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK036628.1 | - | 1 | 2285 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

9130401L11Rik| Gene Symbol: 9130401L11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK036628.1 | - | 1 | 2285 | ||