siRNA Details
Gene Symbol: 9130401L11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK036628.1 | - | 1 | 2026 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BU923475.1 | - | 1 | 344 | Chr. 6 - 98969511 - 98970110 [-] on Build GRCm38 |
