siRNA Details
| Gene Symbol: 8030423F21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045738.1 | 1 | 21 | |||
| GenBank mRNA | AK033101.1 | - | 1 | 22 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB624154.1 | - | 1 | 22 | Chr. 5 - 52609735 - 52619011 [-] on Build GRCm38 | |

8030423F21Rik
Genomic Map