siRNA Details
| Gene Symbol: 2900082C11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK013809.1 | - | 1 | 225 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY712771.1 | - | 1 | 223 | Chr. 12 - 117531831 - 117532293 [+] on Build GRCm38 | |

2900082C11Rik
Genomic Map