siRNA Details
| Gene Symbol: Dcaf12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017944.1 | - | 1 | 390 | ||
| AK083369.1 | - | 1 | 2158 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dcaf12| Gene Symbol: Dcaf12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017944.1 | - | 1 | 390 | ||
| AK083369.1 | - | 1 | 2158 | |||