siRNA Details
| Gene Symbol: Gm30332 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK146614.1 | - | 3 | 469 | ||
| AK169220.1 | - | 3 | 469 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm30332| Gene Symbol: Gm30332 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK146614.1 | - | 3 | 469 | ||
| AK169220.1 | - | 3 | 469 | |||