siRNA Details
| Gene Symbol: Gm10431 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK154292.1 | - | 1 | 2427 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm10431| Gene Symbol: Gm10431 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK154292.1 | - | 1 | 2427 | ||