siRNA Details
| Gene Symbol: D830032E09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102306.1 | 4 | 432 | |||
| NR_102307.1 | 5 | 546 | ||||
| GenBank mRNA | AK085905.1 | - | 4 | 433 | ||
| AK085943.1 | - | 5 | 521 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

D830032E09Rik| Gene Symbol: D830032E09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102306.1 | 4 | 432 | |||
| NR_102307.1 | 5 | 546 | ||||
| GenBank mRNA | AK085905.1 | - | 4 | 433 | ||
| AK085943.1 | - | 5 | 521 | |||