siRNA Details
Gene Symbol: D830032E09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_102306.1 | 4 | 2025 | |||
NR_102307.1 | 5 | 2139 | ||||
GenBank mRNA | AK085905.1 | - | 4 | 2026 | ||
AK085943.1 | - | 5 | 2114 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: D830032E09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_102306.1 | 4 | 2025 | |||
NR_102307.1 | 5 | 2139 | ||||
GenBank mRNA | AK085905.1 | - | 4 | 2026 | ||
AK085943.1 | - | 5 | 2114 |