siRNA Details
| Gene Symbol: A630010A05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033556.1 | 1 | 129 | |||
| GenBank mRNA | AK041445.1 | - | 1 | 130 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB638120.1 | - | 1 | 127 | Chr. 16 - 14562319 - 14562662 [+] on Build GRCm38 | |

A630010A05Rik
Genomic Map