siRNA Details
| Gene Symbol: A630010A05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033556.1 | 6 | 1142 | |||
| GenBank mRNA | AK041445.1 | - | 6 | 1143 | ||
| AK139693.1 | - | 4 | 619 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB228179.2 | - | 3 | 325 | Chr. 16 - 14584331 - 14609814 [+] on Build GRCm38 | |

A630010A05Rik
Genomic Map