siRNA Details
| Gene Symbol: A630010A05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033556.1 | 3 | 403 | |||
| GenBank mRNA | AK041445.1 | - | 3 | 404 | ||
| AK139693.1 | - | 2 | 338 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A630010A05Rik| Gene Symbol: A630010A05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033556.1 | 3 | 403 | |||
| GenBank mRNA | AK041445.1 | - | 3 | 404 | ||
| AK139693.1 | - | 2 | 338 | |||