siRNA Details
| Gene Symbol: A630010A05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033556.1 | 2 | 366 | |||
| GenBank mRNA | AK041445.1 | - | 2 | 367 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A630010A05Rik| Gene Symbol: A630010A05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033556.1 | 2 | 366 | |||
| GenBank mRNA | AK041445.1 | - | 2 | 367 | ||