siRNA Details
| Gene Symbol: 4930480K23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK133329.1 | - | 5 | 1196 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CN528258.1 | - | 4 | 688 | Chr. 14 - 69738238 - 69743818 [+] on Build GRCm38 | |

4930480K23Rik
Genomic Map