siRNA Details
| Gene Symbol: 1600019K03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040481.1 | 3 | 356 | |||
| GenBank mRNA | AK005497.1 | - | 3 | 356 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1600019K03Rik| Gene Symbol: 1600019K03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040481.1 | 3 | 356 | |||
| GenBank mRNA | AK005497.1 | - | 3 | 356 | ||