siRNA Details
| Gene Symbol: 1600019K03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040481.1 | 2 | 218 | |||
| GenBank mRNA | AK005497.1 | - | 2 | 218 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY705699.1 | - | 2 | 220 | Chr. 16 - 35503170 - 35509419 [-] on Build GRCm38 | |

1600019K03Rik
Genomic Map