siRNA Details
| Gene Symbol: Gm19395 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038015.1 | 1 | 88 | |||
| GenBank mRNA | AK076832.1 | - | 1 | 78 | ||
| BC048600.1 | - | 1 | 74 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm19395| Gene Symbol: Gm19395 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038015.1 | 1 | 88 | |||
| GenBank mRNA | AK076832.1 | - | 1 | 78 | ||
| BC048600.1 | - | 1 | 74 | |||