siRNA Details
| Gene Symbol: Ttll4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK014557.1 | - | 372 | |||
| AK019866.1 | - | 1 | 1706 | |||
| AK129075.1 | - | 18 | 6229 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ttll4| Gene Symbol: Ttll4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK014557.1 | - | 372 | |||
| AK019866.1 | - | 1 | 1706 | |||
| AK129075.1 | - | 18 | 6229 | |||