siRNA Details
| Gene Symbol: Gm5089 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK047660.1 | - | 4 | 356 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB647616.1 | - | 4 | 356 | Chr. 14 - 122365877 - 122406743 [-] on Build GRCm38 | |

Gm5089
Genomic Map