siRNA Details
| Gene Symbol: 3110005L24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AA611589.1 | - | 1 | 288 | ||
| AK014001.1 | - | 1 | 964 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AI594411.1 | - | 1 | 300 | Chr. 9 - 50661195 - 50661630 [+] on Build GRCm38 | |

3110005L24Rik
Genomic Map