siRNA Details
| Gene Symbol: Gm16894 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037980.1 | 1 | 185 | |||
| GenBank mRNA | AK041998.1 | - | 1 | 187 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB638745.1 | - | 1 | 187 | Chr. 1 - 40076394 - 40085853 [-] on Build GRCm38 | |

Gm16894
Genomic Map