siRNA Details
| Gene Symbol: 4933427I22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045159.1 | 2 | 144 | |||
| GenBank mRNA | AK016955.1 | - | 2 | 145 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY716636.1 | - | 2 | 143 | Chr. 4 - 139926815 - 139943518 [-] on Build GRCm38 | |

4933427I22Rik
Genomic Map