siRNA Details
| Gene Symbol: 4933427J07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016956.1 | - | 1 | 837 | ||
| Gene Symbol: Gm10417 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK149882.1 | - | 1 | 277 | ||

4933427J07Rik
Genomic Map