siRNA Details
| Gene Symbol: 4930425K10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038182.1 | 1 | 98 | |||
| GenBank mRNA | AK015200.1 | - | 1 | 100 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY714753.1 | - | 1 | 100 | Chr. 5 - 67933646 - 67943081 [-] on Build GRCm38 | |

4930425K10Rik
Genomic Map