siRNA Details
Gene Symbol: 4930425O10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040545.1 | 4 | 661 | |||
GenBank mRNA | AK015203.1 | - | 4 | 661 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4930425O10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040545.1 | 4 | 661 | |||
GenBank mRNA | AK015203.1 | - | 4 | 661 |