siRNA Details
Gene Symbol: 4930425O10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040545.1 | 1 | 40 | |||
GenBank mRNA | AK015203.1 | - | 1 | 40 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY714758.1 | - | 1 | 40 | Chr. 3 - 138712293 - 138726984 [-] on Build GRCm38 |
