siRNA Details
| Gene Symbol: 4930425P05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015204.1 | - | 1 | 325 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY714759.1 | - | 1 | 325 | Chr. 14 - 32012173 - 32012942 [-] on Build GRCm38 | |

4930425P05Rik
Genomic Map