siRNA Details
| Gene Symbol: 4930426I24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015206.1 | - | 1 | 194 | ||
| AK020166.1 | - | 1 | 522 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CF198157.1 | - | 1 | 117 | Chr. 12 - 76402738 - 76403578 [-] on Build GRCm38 | |

4930426I24Rik
Genomic Map