siRNA Details
| Gene Symbol: Ccdc42os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015975.1 | - | 1 | 974 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc42os| Gene Symbol: Ccdc42os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015975.1 | - | 1 | 974 | ||