siRNA Details
| Gene Symbol: 2610304O13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK011983.1 | - | 1 | 229 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY711177.1 | - | 1 | 229 | Chr. 13 - 28725904 - 28726348 [-] on Build GRCm38 | |

2610304O13Rik
Genomic Map