siRNA Details
| Gene Symbol: 4930597A21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016402.1 | - | 3 | 363 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY716054.1 | - | 3 | 363 | Chr. 11 - 44586317 - 44591229 [+] on Build GRCm38 | |

4930597A21Rik
Genomic Map