siRNA Details
| Gene Symbol: 2810404M03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045497.1 | 6 | 1075 | |||
| GenBank mRNA | AK012985.1 | - | 6 | 1074 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2810404M03Rik| Gene Symbol: 2810404M03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045497.1 | 6 | 1075 | |||
| GenBank mRNA | AK012985.1 | - | 6 | 1074 | ||