siRNA Details
Gene Symbol: 2810404M03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045497.1 | 4, 5 | 430 | |||
GenBank mRNA | AK012985.1 | - | 4, 5 | 430 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY712057.1 | - | 430 | Chr. - - [-] on Build |
