siRNA Details
| Gene Symbol: 2810404M03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045497.1 | 4 | 359 | |||
| GenBank mRNA | AK012985.1 | - | 4 | 359 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY712057.1 | - | 359 | Chr. - - [-] on Build | ||

2810404M03Rik
Genomic Map