siRNA Details
| Gene Symbol: 5330416C01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK030468.1 | - | 1, 2 | 87 | ||
| AK045624.1 | - | 1, 2 | 119 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB644122.1 | - | 1, 2 | 119 | Chr. 13 - 97743914 - 97747411 [-] on Build GRCm38 | |

5330416C01Rik
Genomic Map