siRNA Details
| Gene Symbol: Gm16325 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045949.1 | 4 | 1279 | |||
| GenBank mRNA | AK139061.1 | - | 4 | 1280 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm16325| Gene Symbol: Gm16325 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045949.1 | 4 | 1279 | |||
| GenBank mRNA | AK139061.1 | - | 4 | 1280 | ||