siRNA Details
| Gene Symbol: 2900024D18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK013581.1 | - | 1 | 284 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY712542.1 | - | 1 | 284 | Chr. 13 - 6925283 - 6925851 [-] on Build GRCm38 | |

2900024D18Rik
Genomic Map