siRNA Details
| Gene Symbol: Gm16010 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK085341.1 | - | 159 | |||
| AK142507.1 | - | 365 | ||||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB660951.1 | - | 1 | 159 | Chr. 9 - 97016173 - 97018380 [+] on Build GRCm38 | |

Gm16010
Genomic Map