siRNA Details
| Gene Symbol: Gm16010 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK085341.1 | - | 2912 | |||
| AK142507.1 | - | 3130 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm16010| Gene Symbol: Gm16010 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK085341.1 | - | 2912 | |||
| AK142507.1 | - | 3130 | ||||