siRNA Details
| Gene Symbol: 2900024I21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK013589.1 | - | 1 | 909 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2900024I21Rik| Gene Symbol: 2900024I21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK013589.1 | - | 1 | 909 | ||