siRNA Details
| Gene Symbol: 5730406G12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017502.1 | - | 1 | 790 | ||
| AK086338.1 | - | 1 | 1479 | |||
| BB524769.2 | - | 1 | 181 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

5730406G12Rik| Gene Symbol: 5730406G12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017502.1 | - | 1 | 790 | ||
| AK086338.1 | - | 1 | 1479 | |||
| BB524769.2 | - | 1 | 181 | |||