siRNA Details
Gene Symbol: Gm14243 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY512933.1 | - | 3 | 349 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CA533206.1 | - | 3 | 349 | Chr. 2 - 161179064 - 161214902 [+] on Build GRCm38 |
