siRNA Details
| Gene Symbol: Gm14243 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY512933.1 | - | 1 | 131 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CA533206.1 | - | 1 | 131 | Chr. 2 - 161179064 - 161214902 [+] on Build GRCm38 | |

Gm14243
Genomic Map