siRNA Details
| Gene Symbol: Gm14243 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY512933.1 | - | 3 | 1720 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm14243| Gene Symbol: Gm14243 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY512933.1 | - | 3 | 1720 | ||