siRNA Details
| Gene Symbol: Gm20324 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045068.1 | 2, 3 | 543 | |||
| GenBank mRNA | AK080278.1 | - | 2, 3 | 545 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm20324| Gene Symbol: Gm20324 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045068.1 | 2, 3 | 543 | |||
| GenBank mRNA | AK080278.1 | - | 2, 3 | 545 | ||