siRNA Details
| Gene Symbol: Gm20324 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045068.1 | 3 | 613 | |||
| GenBank mRNA | AK080278.1 | - | 3 | 615 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AA175710.1 | - | 1 | 36 | Chr. 15 - 82901750 - 82905518 [+] on Build GRCm38 | |

Gm20324
Genomic Map