siRNA Details
| Gene Symbol: Gm16508 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY512936.1 | - | 2 | 1657 | ||
| AY512943.1 | - | 2 | 1658 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm16508| Gene Symbol: Gm16508 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY512936.1 | - | 2 | 1657 | ||
| AY512943.1 | - | 2 | 1658 | |||