siRNA Details
| Gene Symbol: 0610031O16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045760.1 | 3 | 726 | |||
| GenBank mRNA | AK002733.1 | - | 3 | 726 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

0610031O16Rik| Gene Symbol: 0610031O16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045760.1 | 3 | 726 | |||
| GenBank mRNA | AK002733.1 | - | 3 | 726 | ||