siRNA Details
| Gene Symbol: 0610031O16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045760.1 | 3 | 651 | |||
| GenBank mRNA | AK002733.1 | - | 3 | 651 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY703239.1 | - | 3 | 651 | Chr. 3 - 138210716 - 138239663 [-] on Build GRCm38 | |

0610031O16Rik
Genomic Map