siRNA Details
| Gene Symbol: Mxd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK013062.1 | - | 1 | 203 | ||
| BC025016.1 | - | 1 | 438 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mxd4| Gene Symbol: Mxd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK013062.1 | - | 1 | 203 | ||
| BC025016.1 | - | 1 | 438 | |||